3,5-Difluoro-o-phenylenediamine ≥98%
Supplier: Matrix Scientific
007008-1G
007008-5G
007008-25G
101791-808EA
60
USD
101791-808
101791-810
101791-812
3,5-Difluoro-o-phenylenediamine ≥98%
3,5-Difluoro-o-phenylenediamine
Hazard code(s): Irritant, Refrigerate
Formula:
C₆H₆F₂N₂ Melting Pt: 48…49 °C |
MDL Number:
MFCD00973899 CAS Number: 2369-29-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...