3,5-Difluoro-o-phenylenediamine ≥98%

Supplier: Matrix Scientific

007008-1G 007008-5G 007008-25G
101791-808EA 60 USD
101791-808 101791-810 101791-812
3,5-Difluoro-o-phenylenediamine ≥98%
3,5-Difluoro-o-phenylenediamine

Hazard code(s): Irritant, Refrigerate

Formula: C₆H₆F₂N₂
Melting Pt: 48…49 °C
MDL Number: MFCD00973899
CAS Number: 2369-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR