2-(Methylsulfonyl)benzoic acid

Supplier: Matrix Scientific

006876-1G 006876-5G
101791-424EA 32.31 USD
101791-424 101791-426
2-(Methylsulfonyl)benzoic acid
2-(Methylsulphonyl)benzoic acid

Hazard: Irritant

Formula: C₈H₈O₄S
Melting Pt: 137…140 °C
MDL Number: MFCD00216492
CAS Number: 33963-55-2

Order Now

Specification Test Results

Melting point 137-140°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR