5-Bromo-2-hydroxypyrimidine ≥97%

Supplier: Matrix Scientific

006767-5G 006767-25G
101791-056EA 27.69 USD
101791-056 101791-058
5-Bromo-2-hydroxypyrimidine ≥97%
5-Bromo-2-hydroxypyrimidine

Hazard code(s): Irritant

Formula: C₄H₃BrN₂O
MDL Number: MFCD01318956
CAS Number: 38353-06-9
EINECS: 253-896-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR