5-Bromo-2-hydroxypyrimidine ≥97%
Supplier: Matrix Scientific
006767-5G
006767-25G
101791-056EA
27.69
USD
101791-056
101791-058
5-Bromo-2-hydroxypyrimidine ≥97%
5-Bromo-2-hydroxypyrimidine
Hazard code(s): Irritant
Formula:
C₄H₃BrN₂O |
MDL Number:
MFCD01318956 CAS Number: 38353-06-9 EINECS: 253-896-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...