4-Iodobenzonitrile ≥98% for synthesis
Supplier: Matrix Scientific
Synonyms:
p-Iodobenzonitrile
006690-25G
006690-100G
101790-924EA
53.85
USD
101790-924
101790-926
4-Iodobenzonitrile ≥98% for synthesis
4-Iodobenzonitrile
Hazard code(s): Irritant
Formula:
C₇H₄IN MW: 229.02 g/mol Boiling Pt: 285 Melting Pt: 126…128 °C |
MDL Number:
MFCD00051310 CAS Number: 3058-39-7 UN: 3439 ADR: 6.1 III |
Specification Test Results
Melting point | 126-128°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...