1H,1H-Pentafluoropropyl methacrylate ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: 1H,1H-Pentafluoropropyl methacrylate, 2,2,3,3,3-Pentafluoropropyl Methacrylate, Methacrylic acid 2,2,2,3,3-pentafluoropropyl ester, Methacrylic acid 2,2,3,3,3-Pentafluoropropyl ester, 1H,1H-Perfluoropropyl methacrylate

006275-5G 006275-25G
101790-314EA 61.54 USD
101790-314 101790-316
1H,1H-Pentafluoropropyl methacrylate ≥97% for synthesis
1H,1H-Pentafluoropropyl methacrylate

Hazard code(s): Irritant, Refrigerate

Formula: C₇H₇F₅O₂
MW: 218.12
Boiling Pt: 54…55 °C (100mm)
Density: 1.277 g/cm³
MDL Number: MFCD00039256
CAS Number: 45115-53-5
EINECS: 256-191-8
UN: 3272
ADR: 3 III

Order Now

Specification Test Results

Boiling point 55°C/100mm
Density 1.573
ND20 1.344


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR