Chloropentafluorobenzene ≥99% for synthesis
Supplier: Matrix Scientific
Synonyms:
Pentafluorochlorobenzene
006261-25G
006261-100G
006261-500G
101790-286EA
36.92
USD
101790-286
101790-288
101790-290
Chloropentafluorobenzene ≥99% for synthesis
Chloropentafluorobenzene
Hazard code(s): Irritant
Formula:
C₆ClF₅ Boiling Pt: 122…123 °C (750mm) Melting Pt: –15 °C Density: 1.568 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00000534 CAS Number: 344-07-0 EINECS: 206-450-6 |
Specification Test Results
Boiling point | 122-123°C/7 |
Density | 1.568 |
ND20 | 1.423 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...