5-(4-Fluorophenyl)cyclohexane-1,3-dione, Technical Grade

Supplier: Matrix Scientific

006088-250MG 006088-1G
101790-036EA 44.62 USD
101790-036 101790-038
5-(4-Fluorophenyl)cyclohexane-1,3-dione, Technical Grade
5-(4-Fluorophenyl)cyclohexane-1,3-dione

Hazard: Irritant

Formula: C₁₂H₁₁FO₂
Melting Pt: 183…187 °C
MDL Number: MFCD00174037
CAS Number: 55579-72-1

Order Now

Specification Test Results

Melting point 183-187°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR