5,7-Bis(trifluoromethyl)-4-chloroquinoline ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-5,7-bis(trifluoromethyl)quinoline

005815-250MG 005815-1G
101789-578EA 127.69 USD
101789-578 101789-580
5,7-Bis(trifluoromethyl)-4-chloroquinoline ≥98%
5,7-Bis(trifluoromethyl)-4-chloroquinoline

Hazard: Irritant

Formula: C₁₁H₄ClF₆N
MW: 299.6 g/mol
CAS Number: 234450-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR