Methyl perfluorohexadecanoate ≥95%
Supplier: Matrix Scientific
005783-5G
005783-25G
005783-100G
101789-510EA
53.85
USD
101789-510
101789-512
101789-514
Methyl perfluorohexadecanoate ≥95%
Methyl perfluorohexadecanoate
Hazard: Flammable
Formula:
C₁₇H₃F₃₁O₂ Boiling Pt: 167 °C (50 mmHg) |
MDL Number:
MFCD00236617 CAS Number: 165457-57-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...