(4-Phenoxyphenyl)acetic acid ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 4-Phenoxyphenylacetic Acid

005710-1G 005710-5G
101789-400EA 36.92 USD
101789-400 101789-402
(4-Phenoxyphenyl)acetic acid ≥98% for synthesis
(4-Phenoxyphenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₅OC₆H₅)CH₂COOH
Melting Pt: 79…81 °C
MDL Number: MFCD00079779
CAS Number: 6328-74-1
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Melting point 79-81°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR