α,α,α-Trifluoro-p-toluic acid ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 4-(Trifluoromethyl)benzoic acid

005478-100G
101789-024EA 61.54 USD
101789-024
α,α,α-Trifluoro-p-toluic acid ≥98%
α,α,α-Trifluoro-p-toluic acid

Hazard code(s): Irritant

Formula: C₈H₅F₃O₂
MW: 190.12 g/mol
Boiling Pt: 247 °C (753mm)
Melting Pt: 219…220 °C
MDL Number: MFCD00002562
CAS Number: 455-24-3
EINECS: 207-242-8

Order Now

Specification Test Results

Boiling point 247°C/753mm
Melting point 219-220°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR