3-(Trifluoromethoxy)-DL-phenylglycine

Supplier: Matrix Scientific

005341-1G
101788-690EA 169.23 USD
101788-690
3-(Trifluoromethoxy)-DL-phenylglycine
3-(Trifluoromethoxy)-DL-phenylglycine

Hazard: Irritant

Formula: C₉H₈F₃NO₃
MW: 235.16 g/mol
MDL Number: MFCD01631460
CAS Number: 261952-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR