Bis(pinacolato)diboron ≥98%

Supplier: Matrix Scientific

Synonyms: 4,4,4',4',5,5,5',5'-Octamethyl-2,2'-bi(1,3,2-dioxaborolane), 4,4,5,5-tetramethyl-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane

004889-25G
101787-966EA 27.69 USD
101787-966
Bis(pinacolato)diboron ≥98%
Bis(pinacolato)diboron

Hazard: Irritant/Irritant-Harmful/Refrigerate

Formula: C₁₂H₂₄B₂O₄
MW: 253.94 g/mol
Melting Pt: ∼ 135 °C
MDL Number: MFCD00799570
CAS Number: 73183-34-3

Order Now

Specification Test Results

Melting point ca 135°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR