5-Fluoroindole ≥96% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 5-Fluoro-1H-indole

004607-5G 004607-25G 004607-100G
101787-572EA 80 USD
101787-572 101787-574 101787-576
5-Fluoroindole ≥96% for synthesis
5-Fluoroindole

Hazard code(s): Irritant, Store in Freezer

Formula: C₈H₆FN
Boiling Pt: 85 °C (0.5 mmHg)
Melting Pt: 44…46 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005671
CAS Number: 399-52-0
EINECS: 206-917-4

Order Now

Specification Test Results

Melting point 44-46°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR