N-(tert-Butoxycarbonyl)-4-fluoro-D-phenylalanine
Supplier: Matrix Scientific
Synonyms:
Boc-D-Phe(4-F)-OH, N-Boc-4-fluoro-D-phenylalanine
004417-1G
101787-208EA
27.69
USD
101787-208
N-(tert-Butoxycarbonyl)-4-fluoro-D-phenylalanine
N-(tert-Butoxycarbonyl)-4-fluoro-D-phenylalanine
CAS: 57292-45-2
MDL Number: MFCD00079669
Molecular Formula: C14H18FNO4
Molecular Weight: 283,3
Hazard: Irritant
Formula:
C₁₄H₁₈FNO₄ Melting Pt: 83…86 °C |
MDL Number:
MFCD00079669 CAS Number: 57292-45-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...