1-(4-(Trifluoromethyl)phenyl)imidazole ≥98%

Supplier: Matrix Scientific

004413-1G
101787-204EA 126.15 USD
101787-204
1-(4-(Trifluoromethyl)phenyl)imidazole ≥98%
1-(4-(Trifluoromethyl)phenyl)imidazole

Hazard code(s): Irritant

Formula: C₁₀H₇F₃N₂
Boiling Pt: 90 °C (0.05mm)
Melting Pt: 71…73 °C
MDL Number: MFCD00060493
CAS Number: 25371-98-6

Order Now

Specification Test Results

Boiling point 90°C/0.05mm
Melting point 71-73°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR