2,2'-Bithiophene ≥97%

Supplier: Matrix Scientific

004200-25G
101786-934EA 75.38 USD
101786-934
2,2'-Bithiophene ≥97%
2,2'-Bithiophene

Hazard code(s): Irritant

Formula: C₈H₆S₂
Boiling Pt: 260 °C
Melting Pt: 32…33 °C
MDL Number: MFCD00005414
CAS Number: 492-97-7
EINECS: 207-767-2

Order Now

Specification Test Results

Boiling point 260°C
Melting point 32-33°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR