Ethyl-2-chloro-4,4,4-trifluoroacetoacetate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-chloro-4,4,4-trifluoro-3-oxobutanoate

003979-5G 003979-25G
101786-558EA 44.62 USD
101786-558 101786-560
Ethyl-2-chloro-4,4,4-trifluoroacetoacetate ≥95%
Ethyl-2-chloro-4,4,4-trifluoroacetoacetate

Hazard: Corrosive/Lachrymator

Formula: C₆H₆ClF₃O₃
Boiling Pt: 67…71 °C (35 mmHg)
Flash Pt: 67 °C
MDL Number: MFCD00041540
CAS Number: 363-58-6

Order Now

Specification Test Results

Boiling point 67-71°C/35m
ND20 1.388


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR