2-Chlorothioanisole ≥99%

Supplier: Matrix Scientific
Warning

003953-5G 003953-25G 003953-100G
101786-514EA 20 USD
101786-514 101786-516 101786-518
2-Chlorothioanisole ≥99%
2-Chlorothioanisole

Hazard code(s): Stench

Formula: C₇H₇ClS
Boiling Pt: 116…117 °C (15mm)
MDL Number: MFCD00040124
CAS Number: 17733-22-1
UN: 3334
ADR: 9,

Order Now

Specification Test Results

Boiling point 116-117°C/1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR