1-[3-(Trifluoromethyl)phenyl]but-1-en-3-one

Supplier: Matrix Scientific

003754-250MG 003754-1G
101786-168EA 35.38 USD
101786-168 101786-170
1-[3-(Trifluoromethyl)phenyl]but-1-en-3-one
1-[3-(Trifluoromethyl)phenyl]but-1-en-3-one

Hazard: Irritant

Formula: C₁₁H₉F₃O
Boiling Pt: 115 °C (2.5 mmHg)
MDL Number: MFCD00052843
CAS Number: 80992-92-3

Order Now

Specification Test Results

Boiling point 115°C/2.5mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR