2,2,3,3,3-Pentafluoropropyl difluoromethyl ether ≥97%
Supplier: Matrix Scientific
Synonyms:
Difluoromethyl 2,2,3,3,3-pentafluoropropyl ether
003377-1G
101785-638EA
203.08
USD
101785-638
2,2,3,3,3-Pentafluoropropyl difluoromethyl ether ≥97%
2,2,3,3,3-Pentafluoropropyl difluoromethyl ether
Hazard: Irritant
Formula:
C₄H₃F₇O Boiling Pt: 46 °C Density: 1.46 |
CAS Number:
56860-81-2 |
Specification Test Results
Boiling point | 46°C |
Density | 1.46 |
ND20 | 1.283 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...