2-Fluorobenzoic acid ≥97% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: o-Fluorobenzoic acid

003291-100G 003291-2.5KG
101785-524EA 44.62 USD
101785-524 101785-528
2-Fluorobenzoic acid ≥97% for synthesis
2-Fluorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₅FO₂
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 123…125 °C
Density: 1.46 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002405
CAS Number: 445-29-4
EINECS: 207-158-1

Order Now

Specification Test Results

Melting point 123-125°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR