4-Iodo-1H-pyrazole ≥99%

Supplier: Matrix Scientific

002999-25G
101785-036EA 27.69 USD
101785-036
4-Iodo-1H-pyrazole ≥99%
4-Iodo-1H-pyrazole

Hazard: Irritant

Formula: C₃H₃IN₂
Melting Pt: 108 °C
MDL Number: MFCD00005244
CAS Number: 3469-69-0

Order Now

Specification Test Results

Melting point 108°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR