N-Boc-4-(trifluoromethyl)-L-phenylalanine
Supplier: Matrix Scientific
Synonyms:
N-(tert-Butoxycarbonyl)-4-trifluoromethyl-L-phenylalanine, Boc-Phe(4-CF3)-OH, N-Boc-4-trifluoromethyl-L-phenylalanine
002881-250MG
002881-1G
101784-836EA
61.54
USD
101784-836
101784-838
N-Boc-4-(trifluoromethyl)-L-phenylalanine
N-Boc-4-(trifluoromethyl)-L-phenylalanine
CAS: 114873-07-3
MDL Number: MFCD00792407
Molecular Formula: C15H18F3NO4
Molecular Weight: 333,31
Hazard: Irritant
Formula:
C₁₅H₁₈F₃NO₄ MW: 333.31 g/mol |
MDL Number:
MFCD00792407 CAS Number: 114873-07-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...