2-(2,2,2-Trifluoroethoxy)pyridine-3-carbonitrile ≥96%

Supplier: Matrix Scientific

Synonyms: 2-(2,2,2-Trifluoroethoxy)nicotinonitrile

002878-250MG 002878-1G
101784-828EA 21.54 USD
101784-828 101784-830
2-(2,2,2-Trifluoroethoxy)pyridine-3-carbonitrile ≥96%
2-(2,2,2-Trifluoroethoxy)pyridine-3-carbonitrile

Purity: 96% CAS: 175277-89-1 MDL Number: MFCD00275529 Molecular Formula: C8H5F3N2O Molecular Weight: 202,14 Hazard: Irritant

Formula: C₈H₅F₃N₂O
Melting Pt: 44…46 °C
MDL Number: MFCD00275529
CAS Number: 175277-89-1

Order Now

Specification Test Results

Melting point 44-46°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR