2',4',5'-Trifluoroacetophenone ≥99% for synthesis

Supplier: Matrix Scientific

Synonyms: 1-(2,4,5-Trifluorophenyl)ethan-1-one, 2,4,5-Trifluoroacetophenone

002877-5G 002877-25G
101784-824EA 36.92 USD
101784-824 101784-826
2',4',5'-Trifluoroacetophenone ≥99% for synthesis
2',4',5'-Trifluoroacetophenone

Hazard code(s): Irritant

Formula: C₈H₅F₃O
Boiling Pt: 76…79 °C
Melting Pt: –7 °C
Density: 1.331 g/cm³
MDL Number: MFCD00061193
CAS Number: 129322-83-4

Order Now

Specification Test Results

Boiling point 76-79°C
Melting point -7°C
Density 1.331
ND20 1.472


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR