6-Fluoropicolinic acid ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-6-pyridinecarboxylic acid, 6-Fluoropyridine-2-carboxylic acid

002799-5G 002799-25G
101784-692EA 60 USD
101784-692 101784-694
6-Fluoropicolinic acid ≥97% for synthesis
6-Fluoropicolinic acid

Hazard code(s): Irritant

Formula: C₆H₄FNO₂
Melting Pt: 135…137 °C
MDL Number: MFCD02181193
CAS Number: 402-69-7

Order Now

Specification Test Results

Melting point 135-137°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR