4-Chloro-6-(trifluoromethyl)quinoline ≥98%

Supplier: Matrix Scientific

002738-1G 002738-5G
101784-552EA 115.38 USD
101784-552 101784-554
4-Chloro-6-(trifluoromethyl)quinoline ≥98%
4-Chloro-6-(trifluoromethyl)quinoline

Hazard: Irritant

Formula: C₁₀H₅ClF₃N
MDL Number: MFCD00153106
CAS Number: 49713-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR