2-Chloro-5-fluorobenzoic acid ≥95% for synthesis

Supplier: Matrix Scientific
Warning

002694-25G
101784-452EA 58.46 USD
101784-452
2-Chloro-5-fluorobenzoic acid ≥95% for synthesis
2-Chloro-5-fluorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄ClFO₂
Melting Pt: 147…149 °C
MDL Number: MFCD00156967
CAS Number: 2252-50-8

Order Now

Specification Test Results

Melting point 147-149°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR