3-Chloro-4-fluorobenzotrifluoride ≥98%

Supplier: Matrix Scientific

002499-5G
101784-006EA 76.92 USD
101784-006
3-Chloro-4-fluorobenzotrifluoride ≥98%
3-Chloro-4-fluorobenzotrifluoride

Hazard code(s): Irritant

Formula: C₇H₃ClF₄
MDL Number: MFCD00042207
CAS Number: 78068-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR