2'-Bromo-4'-fluoroacetophenone ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 1-(2-Bromo-4-fluorophenyl)ethanone

002405-5G
101783-852EA 58.46 USD
101783-852
2'-Bromo-4'-fluoroacetophenone ≥98% for synthesis
2'-Bromo-4'-fluoroacetophenone

Hazard code(s): Irritant

Formula: C₈H₆BrFO
Boiling Pt: 150 °C (12mm)
Melting Pt: 48…50 °C
MDL Number: MFCD00077464
CAS Number: 1006-39-9

Order Now

Specification Test Results

Boiling point 150°C/12mm
Melting point 48-50°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR