4-Chloro-3-trifluoromethyl-5-(methyl)pyrazole ≥97%

Supplier: Matrix Scientific

002397-1G 002397-5G
101783-834EA 24.62 USD
101783-834 101783-836
4-Chloro-3-trifluoromethyl-5-(methyl)pyrazole ≥97%
4-Chloro-3-trifluoromethyl-5-(methyl)pyrazole

Hazard: Irritant

Formula: C₅H₄ClF₃N₂
MDL Number: MFCD00155724
CAS Number: 235106-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR