1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-Heptadecafluoro-10-iododecane ≥97% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 1H,1H,2H,2H-Perfluoro-1-iododecane, 1H,1H,2H,2H-Perfluorodecyliodide, 1-Iodo-1H,1H,2H,2H-perfluorodecane

002226-5G
101783-500EA 30.77 USD
101783-500
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-Heptadecafluoro-10-iododecane ≥97% for synthesis
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-Heptadecafluoro-10-iododecane

Hazard code(s): Irritant-Harmful

Formula: C₁₀H₄F₁₇I
Boiling Pt: 178…180 °C
Melting Pt: 53…57 °C
Density: 1.88 g/cm³ (60 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039411
CAS Number: 2043-53-0
EINECS: 218-053-5

Order Now

Specification Test Results

Boiling point 178-180°C
Melting point 53-57°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR