α,α,α,4-Tetrafluoro-o-toluic acid ≥97% for synthesis
Supplier: Matrix Scientific
001976-25G
001976-100G
101782-976EA
56.92
USD
101782-976
101782-978
α,α,α,4-Tetrafluoro-o-toluic acid ≥97% for synthesis
α,α,α,4-Tetrafluoro-o-toluic acid
Hazard code(s): Irritant
Formula:
C₈H₄F₄O₂ MW: 208.11 Boiling Pt: 251 Melting Pt: 121…124 °C |
MDL Number:
MFCD00040982 CAS Number: 141179-72-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...