4-Nitro-2-(trifluoromethyl)aniline ≥98%

Supplier: Matrix Scientific
Warning

001845-25G 001845-100G
101782-712EA 33.85 USD
101782-712 101782-714
4-Nitro-2-(trifluoromethyl)aniline ≥98%
4-Nitro-2-(trifluoromethyl)aniline

Hazard code(s): Irritant

Formula: C₇H₅F₃N₂O₂
Melting Pt: 90…92 °C
Density: 1.754 g/cm³ (–123.15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007365
CAS Number: 121-01-7
EINECS: 204-443-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 90-92°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR