5-Amino-3-methylisoxazole ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 3-Methylisoxazol-5-ylamine, 3-Methylisoxazol-5-amine

001829-5G
101782-682EA 127.69 USD
101782-682
5-Amino-3-methylisoxazole ≥98% for synthesis
5-Amino-3-methylisoxazole

Hazard code(s): Refrigerate

Formula: C₄H₆N₂O
MW: 98.1 g/mol
Boiling Pt: 194
Melting Pt: 84
MDL Number: MFCD00003151
CAS Number: 14678-02-5
EINECS: 238-719-9

Order Now

Specification Test Results

Melting point 81-83°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR