5-Bromo-2-iodotoluene ≥99%

Supplier: Matrix Scientific

001783-5G
101782-568EA 36.92 USD
101782-568
5-Bromo-2-iodotoluene ≥99%
5-Bromo-2-iodotoluene

Hazard code(s): Irritant

Formula: C₇H₆BrI
Boiling Pt: 140 °C (22mm)
Density: 2.08 g/cm³
MDL Number: MFCD00060664
CAS Number: 116632-39-4

Order Now

Specification Test Results

Boiling point 140°C/22mm
Density 2.08


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR