tert-Butyl-4-fluorobenzoate ≥96%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-fluorobenzoate

001730-5G 001730-25G
101782-426EA 41.54 USD
101782-426 101782-428
tert-Butyl-4-fluorobenzoate ≥96%
tert-Butyl 4-fluorobenzoate

Hazard: Irritant

Formula: C₁₁H₁₃FO₂
Boiling Pt: 51…52 °C (0.4 mmHg)
MDL Number: MFCD01320759
CAS Number: 58656-98-7

Order Now

Specification Test Results

Boiling point 51-52°C/0.4


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR