2-Amino-5-(trifluoromethyl)-1,3,4-thiadiazole ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-trifluoromethyl-1,3,4-thiadiazole

001339-1G 001339-5G
101781-430EA 20 USD
101781-430 101781-432
2-Amino-5-(trifluoromethyl)-1,3,4-thiadiazole ≥98% for synthesis
2-Amino-5-(trifluoromethyl)-1,3,4-thiadiazole

Hazard code(s): Irritant

Formula: C₃H₂F₃N₃S
Melting Pt: 225…227 °C
MDL Number: MFCD00003109
CAS Number: 10444-89-0
EINECS: 233-930-2

Order Now

Specification Test Results

Melting point 225-227°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR