1,1,2,2,3,3-Hexafluoropropane ≥97%

Supplier: Matrix Scientific

Synonyms: 1H,3H-Hexafluoropropane

001193-5G
101781-120EA 381.54 USD
101781-120
1,1,2,2,3,3-Hexafluoropropane ≥97%
1,1,2,2,3,3-Hexafluoropropane

Hazard: Nonflammable Gas

Formula: C₃H₂F₆
Boiling Pt: 5 °C
MDL Number: MFCD02093293
CAS Number: 27070-61-7

Order Now

Specification Test Results

Boiling point 5°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR