3',5'-Bis(trifluoromethyl)propiophenone ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 1-(3,5-Bis(trifluoromethyl)phenyl)propan-1-one

001103-1G 001103-5G
101780-962EA 15.38 USD
101780-962 101780-964
3',5'-Bis(trifluoromethyl)propiophenone ≥98% for synthesis
3',5'-Bis(trifluoromethyl)propiophenone

Hazard code(s): Irritant

Formula: C₁₁H₈F₆O
Density: 1.379 g/cm³
MDL Number: MFCD00009911
CAS Number: 85068-34-4
EINECS: 285-296-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Density 1.379
ND20 1.4245


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR