7-Bromo-1-heptene 97%
Supplier: Matrix Scientific
|
Danger
|
054186-5G
101780-650EA
30.77
USD
101780-650
7-Bromo-1-heptene 97%
7-Bromo-1-heptene
Purity: 97%
CAS: 2695-47-8
MDL Number: MFCD01631137
Molecular Formula: C7H13Br
Molecular Weight: 177,09
Boiling Point: 27°C/1mm
TCA listed: T
Hazard: Flammable
Formula:
C₇H₁₃Br Boiling Pt: 27 °C (1 mmHg) Density: 1.2169 (20 °C) |
MDL Number:
MFCD01631137 CAS Number: 4117-09-3 UN: 1993 ADR: 3,III |
Specification Test Results
Boiling point | 27°/1mm |
Density | 1.162 |
ND20 | 1.466 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...