2-Fluoro-5-(trifluoromethyl)phenyl isocyanate ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-5-(trifluoromethyl)phenylisocyanate

032429-25G 032429-1G 032429-5G
101780-532EA 573.85 USD
101780-532 101780-528 101780-530
2-Fluoro-5-(trifluoromethyl)phenyl isocyanate ≥97%
2-Fluoro-5-(trifluoromethyl)phenyl isocyanate

Hazard: Toxic/Irritant-Harmful/Refrigerate/Lachrymator

Formula: C₈H₃F₄NO
Boiling Pt: 53 °C (2.5 mmHg)
MDL Number: MFCD00673069
CAS Number: 69922-27-6
UN: 2922
ADR: 8,III

Order Now

Specification Test Results

Boiling point 53°/2.5mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR