3-Aminobenzonitrile ≥99%

Supplier: Matrix Scientific

012613-100G
101780-128EA 186.15 USD
101780-128
3-Aminobenzonitrile ≥99%
3-Aminobenzonitrile

Hazard: Toxic

Formula: C₇H₆N₂
Boiling Pt: 289 °C
Melting Pt: 51…53 °C
MDL Number: MFCD00007756
CAS Number: 2237-30-1
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 51-53°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR