2-Piperazinone ≥97%
Supplier: Matrix Scientific
Synonyms:
Piperazin-2-one
012421-5G
012421-25G
101780-102EA
30.77
USD
101780-102
101780-104
2-Piperazinone ≥97%
2-Piperazinone
Hazard: Corrosive
Formula:
C₄H₈N₂O MW: 100.12 g/mol Boiling Pt: 165 °C (5 Torr) Melting Pt: 135…137 °C |
MDL Number:
MFCD01318687 CAS Number: 5625-67-2 |
Specification Test Results
Melting point | 135-137° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...