3-Hydroxythiophenol ≥98%
Supplier: Matrix Scientific
Synonyms:
3-Hydroxybenzene-1-thiol, 3-Hydroxybenzenethiol, 3-Hydroxythiophenol, 1-Hydroxy-3-mercaptobenzene, 3-Mercaptophenol, m-Hydroxythiophenol, 3-Hydroxyphenyl mercaptan
011292-5G
011292-25G
101780-088EA
41.54
USD
101780-088
101780-090
3-Hydroxythiophenol ≥98%
3-Hydroxythiophenol
Hazard: Corrosive/Air Sensitive/Stench
Formula:
C₆H₆OS MW: 126.18 Boiling Pt: 168 °C (15 Torr) Melting Pt: 16.5 Density: 1.237 |
MDL Number:
MFCD01317829 CAS Number: 40248-84-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...