2,3,5-Trifluoropyridine ≥95%

Supplier: Matrix Scientific

009192-5G 009192-25G
101779-916EA 166.15 USD
101779-916 101779-918
2,3,5-Trifluoropyridine ≥95%
2,3,5-Trifluoropyridine

Hazard: Flammable

Formula: C₅H₂F₃N
Boiling Pt: 102 °C
Density: 1.499
MDL Number: MFCD03001162
CAS Number: 76469-41-5
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 102°
Density 1.499
ND20 1.422


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR