2-Chlorobenzylamine ≥97%

Supplier: Matrix Scientific

Synonyms: (2-Chlorophenyl)methanamine

007530-25G
101779-592EA 55.38 USD
101779-592
2-Chlorobenzylamine ≥97%
2-Chlorobenzylamine

TCA listed: T
Hazard: Corrosive/Air Sensitive

Formula: C₇H₈ClN
Boiling Pt: 103…104 °C (11 mmHg)
Density: 1.17
MDL Number: MFCD00008108
CAS Number: 89-97-4
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Boiling point 103-104°/1
Density 1.17
ND20 1.563


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR